Propyl-2-furancarboxylate 95%
Supplier: Apollo Scientific
Synonyms:
Propyl 2-Furoate, 2-Furoic Acid Propyl Ester, 2-Furancarboxylic Acid Propyl Ester
OR937748-25G
OR937748-5G
APOSOR937748-25GEA
226
EUR
APOSOR937748-25G
APOSOR937748-5G
Propyl-2-furancarboxylate 95%
Propyl-2-furancarboxylate
Formula:
C₈H₁₀O₃ MW: 154,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00046108 CAS Number: 615-10-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...