6-Methyl-2H-pyrido[1,2-a]pyrimidin-2-one 95%

Supplier: Apollo Scientific

OR937769-10G OR937769-1G OR937769-5G
APOSOR937769-10GEA 935 EUR
APOSOR937769-10G APOSOR937769-1G APOSOR937769-5G
6-Methyl-2H-pyrido[1,2-a]pyrimidin-2-one 95%
6-Methyl-2H-pyrido[1,2-a]pyrimidin-2-one
Formula: C₉H₈N₂O
MW: 160,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191466
CAS Number: 16075-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR