Picolinamide 95%
Supplier: Apollo Scientific
Synonyms:
2-Pyridinecarboxamide, Pyridine-2-carboxamide
OR937776-100G
APOSOR937776-100GEA
130
EUR
APOSOR937776-100G
Picolinamide 95%
Picolinamide
Formula:
C₆H₆N₂O MW: 122,13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00023483 CAS Number: 1452-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...