4-Nitrophenyl acetate 95%
Supplier: Apollo Scientific
Synonyms:
P-Acetoxynitrobenzene, 1-Acetoxy-4-nitrobenzene
OR937811-100G
APOSOR937811-100GEA
105
EUR
APOSOR937811-100G
4-Nitrophenyl acetate 95%
4-Nitrophenyl acetate
Formula:
C₈H₇NO₄ MW: 181,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00007326 CAS Number: 830-03-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...