2,3,6-Trichlorophenol 95%
Supplier: Apollo Scientific
OR937826-1G
OR937826-5G
APOSOR937826-1GEA
234
EUR
APOSOR937826-1G
APOSOR937826-5G
2,3,6-Trichlorophenol 95%
2,3,6-Trichlorophenol
Formula:
C₆H₃Cl₃O MW: 197,45 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002165 CAS Number: 933-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...