2-Isobutyl-4-methylthiazole 95%

OR937864-10G OR937864-1G OR937864-5G
APOSOR937864-10GEA 590 EUR
APOSOR937864-10G APOSOR937864-1G APOSOR937864-5G
2-Isobutyl-4-methylthiazole 95%
2-Isobutyl-4-methylthiazole
Formula: C₈H₁₃NS
MW: 155,2605 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00053138
CAS Number: 61323-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR