4-(Methylthio)butanone 95%

Supplier: Apollo Scientific

OR937881-1G OR937881-25G OR937881-5G
APOSOR937881-1GEA 116 EUR
APOSOR937881-1G APOSOR937881-25G APOSOR937881-5G
4-(Methylthio)butanone 95%
4-(Methylthio)butanone
Formula: C₅H₁₀OS
Storage Temperature: Ambient
MDL Number: MFCD00040029
CAS Number: 34047-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR