Acetyl sulphide 95%
Supplier: Apollo Scientific
Synonyms:
Diacetyl Sulphide
OR937895-10G
OR937895-1G
OR937895-5G
APOSOR937895-10GEA
806
EUR
APOSOR937895-10G
APOSOR937895-1G
APOSOR937895-5G
Acetyl sulphide 95%
Acetyl sulphide
Formula:
C₄H₆O₂S MW: 118,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00008753 CAS Number: 3232-39-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...