2,2,4-Trimethylcyclopentanone 95%

Supplier: Apollo Scientific

OR937966-1G OR937966-5G
APOSOR937966-1GEA 241 EUR
APOSOR937966-1G APOSOR937966-5G
2,2,4-Trimethylcyclopentanone 95%
2,2,4-Trimethylcyclopentanone
Formula: C₈H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD00019312
CAS Number: 28056-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR