1,6-Naphthyridin-2(1H)-one 97%
Supplier: Apollo Scientific
OR938005-1G
OR938005-250MG
APOSOR938005-1GEA
633
EUR
APOSOR938005-1G
APOSOR938005-250MG
1,6-Naphthyridin-2(1H)-one 97%
1,6-Naphthyridin-2(1H)-one
Formula:
C₈H₆N₂O MW: 146,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07781179 CAS Number: 23616-29-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...