(S)-(-)-1-Phenyl-1-propanol 98%

Supplier: Apollo Scientific

Synonyms: (S)-1-phenyl-1-propanol, (S)-(-)-α-Ethylbenzyl alcohol

OR938065-1G
APOSOR938065-1GEA 137 EUR
APOSOR938065-1G
(S)-(-)-1-Phenyl-1-propanol 98%
(S)-(-)-1-Phenyl-1-propanol
Formula: C₉H₁₂O
MW: 136,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066207
CAS Number: 613-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR