2-(Methylamino)pyrimidine 95%

Supplier: Apollo Scientific

Synonyms: 2-Methylaminopyrimidine

OR938076-1G OR938076-5G
APOSOR938076-1GEA 170 EUR
APOSOR938076-1G APOSOR938076-5G
2-(Methylamino)pyrimidine 95%
2-(Methylamino)pyrimidine
Formula: C₅H₇N₃
MW: 109,132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234126
CAS Number: 931-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR