Methyl-4-vinylbenzoate 95%

Supplier: Apollo Scientific

Synonyms: Methyl 4-vinylbenzoate

OR938080-1G OR938080-5G
APOSOR938080-1GEA 79.9 EUR
APOSOR938080-1G APOSOR938080-5G
Methyl-4-vinylbenzoate 95%
Methyl-4-vinylbenzoate
Formula: C₁₀H₁₀O₂
MW: 162.188 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017218
CAS Number: 1076-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR