Neopentylamine

Supplier: Apollo Scientific
Danger

Synonyms: 1-Amino-2,2-dimethylpropane

OR938214-100ML OR938214-25ML
APOSOR938214-100MLEA 720 EUR
APOSOR938214-100ML APOSOR938214-25ML
Neopentylamine
Neopentylamine
Formula: C₅H₁₃N
MW: 87,16 g/mol
Boiling Pt: 79…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00008134
CAS Number: 5813-64-9
UN: 2733
ADR: 3+8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR