2-(2-Methoxyphenyl)acetyl chloride 95%
Supplier: Apollo Scientific
OR938487-100MG
OR938487-250MG
OR938487-500MG
APOSOR938487-100MGEA
190
EUR
APOSOR938487-100MG
APOSOR938487-250MG
APOSOR938487-500MG
2-(2-Methoxyphenyl)acetyl chloride 95%
2-(2-Methoxyphenyl)acetyl chloride
Formula:
C₉H₉ClO₂ Storage Temperature: Ambient |
MDL Number:
MFCD08899364 CAS Number: 28033-63-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...