2-(3-Thienyl)-1,3-dioxolane 95%
Supplier: Apollo Scientific
Synonyms:
2-(Thiophen-3-yl)-1,3-dioxolane, 3-(1,3-Dioxolan-2-yl)thiophene
OR938624-5G
APOSOR938624-5GEA
42.2
EUR
APOSOR938624-5G
2-(3-Thienyl)-1,3-dioxolane 95%
2-(3-Thienyl)-1,3-dioxolane
Formula:
C₇H₈O₂S MW: 156,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06209359 CAS Number: 13250-82-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...