Methyl (methylthio)acetate

Supplier: Apollo Scientific
Warning

Synonyms: (Methylthio)acetic methyl ester

OR9387-100G OR9387-25G OR9387-5G
APOSOR9387-100GEA 277 EUR
APOSOR9387-100G APOSOR9387-25G APOSOR9387-5G
Methyl (methylthio)acetate
Methyl (methylthio)acetate
Formula: C₄H₈O₂S
MW: 120,17 g/mol
Boiling Pt: 49…51 °C/12mmHg
Density: 1,11 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039809
CAS Number: 16630-66-3
EINECS: 240-683-4
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR