3-Bromoindole 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Bromo-1H-indole
OR938836-250MG
OR938836-1G
OR938836-100MG
APOSOR938836-250MGEA
1590
EUR
APOSOR938836-250MG
APOSOR938836-1G
APOSOR938836-100MG
3-Bromoindole 95%
3-Bromoindole
Formula:
C₈H₆BrN MW: 196,05 g/mol Storage Temperature: Refrigerator |
CAS Number:
1484-27-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...