4-Acetylindan 95%
Supplier: Apollo Scientific
OR938861-1G
OR938861-250MG
APOSOR938861-1GEA
538
EUR
APOSOR938861-1G
APOSOR938861-250MG
4-Acetylindan 95%
4-Acetylindan
Formula:
C₁₁H₁₂O Storage Temperature: Ambient |
MDL Number:
MFCD09033715 CAS Number: 38997-97-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...