Methyl-4-isocyanatobenzoate 95%

Supplier: Apollo Scientific

Synonyms: Methyl 4-isocyanatobenzoate

OR938901-1G OR938901-5G
APOSOR938901-1GEA 53.9 EUR
APOSOR938901-1G APOSOR938901-5G
Methyl-4-isocyanatobenzoate 95%
Methyl 4-isocyanatobenzoate

When melted, may contain small amount of sediment.

Formula: C₉H₇NO₃
MW: 177,1568 g/mol
Melting Pt: 50…52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863690
CAS Number: 23138-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR