3-[[tert-Butyl(dimethyl)silyl]oxy]-1-propanol 95%

Supplier: Apollo Scientific

Synonyms: 3-((tert-Butyldimethylsilyl)oxy)propan-1-ol, 3-[[tert-Butyl(dimethyl)silyl]oxy]propan-1-ol, 3-[(tert-Butyldimethylsilyl)oxy]-1-propanol

OR938905-10G OR938905-1G OR938905-5G
APOSOR938905-10GEA 80 EUR
APOSOR938905-10G APOSOR938905-1G APOSOR938905-5G
3-[[tert-Butyl(dimethyl)silyl]oxy]-1-propanol 95%
3-[[tert-Butyl(dimethyl)silyl]oxy]-1-propanol
Formula: C₉H₂₂O₂Si
MW: 190,36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00187968
CAS Number: 73842-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR