2-(4-Chlorophenyl)cyclopropanecarboxylic acid 95%
Supplier: Apollo Scientific
OR938907-1G
OR938907-250MG
APOSOR938907-1GEA
212
EUR
APOSOR938907-1G
APOSOR938907-250MG
2-(4-Chlorophenyl)cyclopropanecarboxylic acid 95%
2-(4-Chlorophenyl)cyclopropanecarboxylic acid
Formula:
C₁₀H₉ClO₂ Storage Temperature: Ambient |
MDL Number:
MFCD03419437 CAS Number: 90940-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...