1-(2-Bromoethyl)pyrrole 97%

Supplier: Apollo Scientific

OR938914-1G
APOSOR938914-1GEA 106 EUR
APOSOR938914-1G
1-(2-Bromoethyl)pyrrole 97%
1-(2-Bromoethyl)pyrrole
Formula: C₆H₈BrN
Storage Temperature: Ambient
MDL Number: MFCD00191322
CAS Number: 78358-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR