1,3-Dihydro-1-methoxyisobenzofuran 95%
Supplier: Apollo Scientific
Synonyms:
1-Methoxyisocoumaran
OR939007-10G
OR939007-1G
OR939007-5G
APOSOR939007-10GEA
1010
EUR
APOSOR939007-10G
APOSOR939007-1G
APOSOR939007-5G
1,3-Dihydro-1-methoxyisobenzofuran 95%
1,3-Dihydro-1-methoxyisobenzofuran
Formula:
C₉H₁₀O₂ MW: 150,18 g/mol Storage Temperature: Ambient |
CAS Number:
67536-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...