1-Methyl-1H-pyrazole-4-carbonitrile 95%

Supplier: Apollo Scientific

Synonyms: 4-Cyano-1-methylpyrazole

OR939081-10G OR939081-1G OR939081-5G
APOSOR939081-10GEA 230 EUR
APOSOR939081-10G APOSOR939081-1G APOSOR939081-5G
1-Methyl-1H-pyrazole-4-carbonitrile 95%
1-Methyl-1H-pyrazole-4-carbonitrile
Formula: C₅H₅N₃
MW: 107.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04969704
CAS Number: 66121-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR