3-bromofuran-2,5-dione 95%
Supplier: Apollo Scientific
Synonyms:
Bromomaleic anhydride
OR939690-10G
OR939690-1G
OR939690-5G
APOSOR939690-10GEA
171
EUR
APOSOR939690-10G
APOSOR939690-1G
APOSOR939690-5G
3-bromofuran-2,5-dione 95%
3-bromofuran-2,5-dione
Formula:
C₄HBrO₃ MW: 176,953 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005519 CAS Number: 5926-51-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...