Diallylamine hydrochloride 95%
Supplier: Apollo Scientific
OR939744-25G
OR939744-5G
APOSOR939744-25GEA
314
EUR
APOSOR939744-25G
APOSOR939744-5G
Diallylamine hydrochloride 95%
Diallylamine hydrochloride
Formula:
C₆H₁₁N Storage Temperature: Ambient |
MDL Number:
MFCD00060150 CAS Number: 6147-66-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...