2,4,6-Trimethylpyridin-3-amine 95%

Supplier: Apollo Scientific

OR939891-1G OR939891-250MG OR939891-500MG
APOSOR939891-1GEA 561 EUR
APOSOR939891-1G APOSOR939891-250MG APOSOR939891-500MG
2,4,6-Trimethylpyridin-3-amine 95%
2,4,6-Trimethylpyridin-3-amine
Formula: C₈H₁₂N₂
Storage Temperature: Ambient
MDL Number: MFCD05257015
CAS Number: 51467-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR