Ethyl Indoline-2-carboxylate 95%
Supplier: Apollo Scientific
Synonyms:
Ethyl indoline-2-carboxylate, Indoline-2-carboxylic Acid Ethyl Ester
OR940768-1G
OR940768-5G
APOSOR940768-1GEA
66.7
EUR
APOSOR940768-1G
APOSOR940768-5G
Ethyl Indoline-2-carboxylate 95%
Ethyl Indoline-2-carboxylate
Formula:
C₁₁H₁₃NO₂ MW: 191,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06797114 CAS Number: 50501-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...