2-Cyano-N,N-diethylacetamide 95%
Supplier: Apollo Scientific
OR941880-1G
OR941880-25G
OR941880-5G
APOSOR941880-1GEA
49.5
EUR
APOSOR941880-1G
APOSOR941880-25G
APOSOR941880-5G
2-Cyano-N,N-diethylacetamide 95%
2-Cyano-N,N-diethylacetamide
Formula:
C₇H₁₂N₂O MW: 140,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01332383 CAS Number: 26391-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...