2-Methyl-4-phenyl-1H-imidazole 95%
Supplier: Apollo Scientific
OR942345-1G
OR942345-5G
APOSOR942345-1GEA
128
EUR
APOSOR942345-1G
APOSOR942345-5G
2-Methyl-4-phenyl-1H-imidazole 95%
2-Methyl-4-phenyl-1H-imidazole
Formula:
C₁₀H₁₀N₂ Storage Temperature: Ambient |
MDL Number:
MFCD01646082 CAS Number: 13739-48-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...