3-Methoxysalicyl alcohol 95%
Supplier: Apollo Scientific
Synonyms:
(2-Hydroxy-3-methoxyphenyl)methanol, 2-(Hydroxymethyl)-6-methoxyphenol, 2-Hydroxy-3-methoxybenzyl alcohol, 2-Hydroxy-3-(hydroxymethyl)anisole
OR943121-25G
OR943121-5G
APOSOR943121-25GEA
148
EUR
APOSOR943121-25G
APOSOR943121-5G
3-Methoxysalicyl alcohol 95%
3-Methoxysalicyl alcohol
Formula:
C₈H₁₀O₃ MW: 154,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00008362 CAS Number: 4383-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...