4-Amino-1-benzylpyrrolidin-2-one 95%

Supplier: Apollo Scientific

OR943174-1G OR943174-250MG
APOSOR943174-1GEA 771 EUR
APOSOR943174-1G APOSOR943174-250MG
4-Amino-1-benzylpyrrolidin-2-one 95%
4-Amino-1-benzylpyrrolidin-2-one
Formula: C₁₁H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD09971803
CAS Number: 368429-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR