(5-Phenyl-1,3,4-oxadiazol-2-yl)methanol 95%
Supplier: Apollo Scientific
OR944559-1G
OR944559-250MG
APOSOR944559-1GEA
648
EUR
APOSOR944559-1G
APOSOR944559-250MG
(5-Phenyl-1,3,4-oxadiazol-2-yl)methanol 95%
(5-Phenyl-1,3,4-oxadiazol-2-yl)methanol
Formula:
C₉H₈N₂O₂ Storage Temperature: Ambient |
CAS Number:
54014-02-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...