4'-Methoxy-3'-methylacetophenone 95%

Supplier: Apollo Scientific

Synonyms: 1-(4-Methoxy-3-methylphenyl)ethanone, 3'-Methyl-4'-methoxyacetophenone

OR944970-5G
APOSOR944970-5GEA 132 EUR
APOSOR944970-5G
4'-Methoxy-3'-methylacetophenone 95%
4'-Methoxy-3'-methylacetophenone
Formula: C₁₀H₁₂O₂
MW: 164.2011 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01673307
CAS Number: 10024-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR