4-Methylnicotinic acid
Supplier: Apollo Scientific
|
Warning
|
OR9450-25G
OR9450-5G
APOSOR9450-25GEA
88.6
EUR
APOSOR9450-25G
APOSOR9450-5G
4-Methylnicotinic acid
4-Methylnicotinic acid
Formula:
C₇H₇NO₂ Melting Pt: 214…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00128869 CAS Number: 3222-50-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...