2-(Hydroxymethyl)phenylboronic acid

Supplier: Apollo Scientific
Warning

OR9454-5G OR9454-1G OR9454-25G
APOSOR9454-5GEA 26.1 EUR
APOSOR9454-5G APOSOR9454-1G APOSOR9454-25G
2-(Hydroxymethyl)phenylboronic acid
2-(Hydroxymethyl)phenylboronic acid
Formula: C₇H₉BO₃
Storage Temperature: Ambient
MDL Number: MFCD02258966
CAS Number: 87199-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR