3-Methyl-3-aminomethylazetidine dihydrochloride

Supplier: Apollo Scientific

Synonyms: (3-Methylazetidin-3-yl)methanamine dihydrochloride

OR945944-1G OR945944-250MG OR945944-500MG
APOSOR945944-1GEA 877 EUR
APOSOR945944-1G APOSOR945944-250MG APOSOR945944-500MG
3-Methyl-3-aminomethylazetidine dihydrochloride
3-Methyl-3-aminomethylazetidine dihydrochloride
Formula: C₅H₁₄Cl₂N₂
MW: 173.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09264501
CAS Number: 96308-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR