4-Propoxybenzaldehyde 96%
Supplier: Apollo Scientific
OR946442-100G
OR946442-25G
APOSOR946442-100GEA
37.3
EUR
APOSOR946442-100G
APOSOR946442-25G
4-Propoxybenzaldehyde 96%
4-Propoxybenzaldehyde
Formula:
C₁₀H₁₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00014134 CAS Number: 5736-85-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...