4-Methyl-1-naphthonitrile 95%
Supplier: Apollo Scientific
Synonyms:
1-Cyano-4-methylnaphthalene
OR946849-1G
OR946849-5G
APOSOR946849-1GEA
36.4
EUR
APOSOR946849-1G
APOSOR946849-5G
4-Methyl-1-naphthonitrile 95%
4-Methyl-1-naphthonitrile
Formula:
C₁₂H₉N MW: 167.2066 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00079742 CAS Number: 36062-93-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...