4-(5-Oxo-3-pyrazolidinyl)benzeneboronic acid 97%
Supplier: Apollo Scientific
|
Warning
|
OR9472-1G
OR9472-250MG
APOSOR9472-1GEA
227
EUR
APOSOR9472-1G
APOSOR9472-250MG
4-(5-Oxo-3-pyrazolidinyl)benzeneboronic acid 97%
4-(5-Oxo-3-pyrazolidinyl)benzeneboronic acid
Formula:
C₉H₁₁BN₂O₃ Melting Pt: 187…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD08235074 CAS Number: 874290-64-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...