4,7-Dimethyl-1H-indole-2,3-dione 95%

OR948082-1G OR948082-250MG
APOSOR948082-1GEA 415 EUR
APOSOR948082-1G APOSOR948082-250MG
4,7-Dimethyl-1H-indole-2,3-dione 95%
4,7-Dimethyl-1H-indole-2,3-dione
Formula: C₁₀H₉NO₂
MW: 175,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047217
CAS Number: 15540-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR