Ethyl trichloroacetate 95%

Supplier: Apollo Scientific

Synonyms: Trichloroacetic acid ethyl ester

OR949648-100G OR949648-25G OR949648-500G
APOSOR949648-100GEA 61.1 EUR
APOSOR949648-100G APOSOR949648-25G APOSOR949648-500G
Ethyl trichloroacetate 95%
Ethyl trichloroacetate
Formula: C₄H₅Cl₃O₂
MW: 191,44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000795
CAS Number: 515-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR