tert-Butyl-3-endo-3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
tert-Butyl 3-endo-3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate
OR950059-1G
OR950059-25G
OR950059-5G
APOSOR950059-1GEA
21.9
EUR
APOSOR950059-1G
APOSOR950059-25G
APOSOR950059-5G
tert-Butyl-3-endo-3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate 97%
tert-Butyl-3-endo-3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate
Formula:
C₁₂H₂₁NO₃ MW: 227,3 g/mol Melting Pt: 141…142,5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD22409620 CAS Number: 143557-91-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...