4-Hexylpyridine 95%

Supplier: Apollo Scientific

OR950306-1G OR950306-250MG
APOSOR950306-1GEA 359 EUR
APOSOR950306-1G APOSOR950306-250MG
4-Hexylpyridine 95%
4-Hexylpyridine
Formula: C₁₁H₁₇N
Storage Temperature: Ambient
MDL Number: MFCD00068555
CAS Number: 27876-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR