Ethyl-2-amino-4-methylpentanoate 95%

Supplier: Apollo Scientific

OR950312-100G OR950312-25G
APOSOR950312-100GEA 261 EUR
APOSOR950312-100G APOSOR950312-25G
Ethyl-2-amino-4-methylpentanoate 95%
Ethyl-2-amino-4-methylpentanoate
Formula: C₈H₁₇NO₂
MW: 159,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00987966
CAS Number: 2899-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR