6-Chloro-2,3-dihydroquinolin-4(1H)-one 95%

Supplier: Apollo Scientific

OR950747-1G OR950747-250MG
APOSOR950747-1GEA 172 EUR
APOSOR950747-1G APOSOR950747-250MG
6-Chloro-2,3-dihydroquinolin-4(1H)-one 95%
6-Chloro-2,3-dihydroquinolin-4(1H)-one
Formula: C₉H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD09834922
CAS Number: 21617-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR