4-Chloronicotinamide 97%

Supplier: Apollo Scientific

Synonyms: 4-Chloropyridine-3-carboxamide

OR951010-1G OR951010-5G
APOSOR951010-1GEA 212 EUR
APOSOR951010-1G APOSOR951010-5G
4-Chloronicotinamide 97%
4-Chloronicotinamide
Formula: C₆H₅ClN₂O
Storage Temperature: Ambient
CAS Number: 7418-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR