Methyl-4-oxocyclohexanecarboxylate 95%

Supplier: Apollo Scientific

Synonyms: Methyl 4-oxocyclohexanecarboxylate, 4-Oxo-cyclohexanecarboxylic acid methyl ester

OR951132-100G
APOSOR951132-100GEA 219 EUR
APOSOR951132-100G
Methyl-4-oxocyclohexanecarboxylate 95%
Methyl-4-oxocyclohexanecarboxylate
Formula: C₈H₁₂O₃
MW: 156,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00270324
CAS Number: 6297-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR