1-Pyridin-2-ylpropan-1-one 97%

Supplier: Apollo Scientific

Synonyms: 2-Propionylpyridine

OR951266-1G OR951266-5G
APOSOR951266-1GEA 119 EUR
APOSOR951266-1G APOSOR951266-5G
1-Pyridin-2-ylpropan-1-one 97%
1-Pyridin-2-ylpropan-1-one
Formula: C₈H₉NO
MW: 135,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00023486
CAS Number: 3238-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR